methyl (2S,3S,4S,5R,6S)-3,4,5-triacetoxy-6-[2-amino-4-(hydroxymethyl)phenoxy]tetrahydropyran-2-carboxylate 1 * 1 g
Catalog # PHBLPBY0198-1G
Supplier: Pharmablock (USA)
methyl (2S,3S,4S,5R,6S)-3,4,5-triacetoxy-6-[2-amino-4-(hydroxymethyl)phenoxy]tetrahydropyran-2-carboxylate 1 * 1 g
Catalog # PHBLPBY0198-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
2,3-Dimercapto-1-propanol for synthesis, Sigma-Aldrich®
Supplier: Merck
2-Propanol, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
m-Xylene for synthesis, Sigma-Aldrich®
Supplier: Merck
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Glycerine (synthetic) 99+%, pure
Supplier: Thermo Fisher Scientific
Benzyl alcohol 99-100.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanolamine, Sigma-Aldrich®
Supplier: Merck
Acetonitrile ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
2-Methylpentane, Sigma-Aldrich®
Supplier: Merck
Turpentine oil
Supplier: Roth Carl
n-Heptane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Cyclohexane for pesticide residue analysis
Supplier: Thermo Fisher Scientific
Ethyl acetate, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
DNACON CALIBRATION SOLUTION 1 * 1 KIT
Supplier: Cole-Parmer