O2-tert-butyl O4-methyl rel-(1S,4R,5S)-2-azabicyclo[3.1.0]hexane-2,4-dicarboxylate 1 * 1 g
Catalog # PHBLPBXY285-1G
Supplier: Pharmablock (USA)
O2-tert-butyl O4-methyl rel-(1S,4R,5S)-2-azabicyclo[3.1.0]hexane-2,4-dicarboxylate 1 * 1 g
Catalog # PHBLPBXY285-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Hydrogen peroxide 35%, J.T.Baker®
Supplier: Avantor
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Toluene, anhydrous (max. 0.005% H₂O) ≥99.8%, AnalaR NORMAPUR® for synthesis
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2,2,4-Trimethylpentane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals