tert-butyl (3R,4S)-3-cyano-3-cyclopropyl-4-methyl-pyrrolidine-1-carboxylate 1 * 1 g
Catalog # PHBLPBX3525-1G
Supplier: Pharmablock (USA)
tert-butyl (3R,4S)-3-cyano-3-cyclopropyl-4-methyl-pyrrolidine-1-carboxylate 1 * 1 g
Catalog # PHBLPBX3525-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
2,2,4-Trimethylpentane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
(±)-2-Butanol ≥99% analytical reagent
Supplier: VWR Chemicals
Tetrahydrofuran, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Chloroform ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Toluene ≥99.9% (by GC), SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Acetone ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Supplier: VWR Chemicals
n-Heptane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
2,2,4-Trimethylpentane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
(±)-2-Butanol, EMSURE® for analysis, Supelco®
Supplier: Merck
tert-Butanol, EMPLURA®, Supelco®
Supplier: Merck
n-Hexane ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Dimethyl sulphoxide 99.8+%, extra pure
Supplier: Thermo Fisher Scientific