3-fluoro-3-methyl-piperidin-4-ol hydrochloride 1 * 1 g
Catalog # PHBLPBX1815-1-1G
Supplier: Pharmablock (USA)
3-fluoro-3-methyl-piperidin-4-ol hydrochloride 1 * 1 g
Catalog # PHBLPBX1815-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Water with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Pyridine, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
Tetrahydrofuran anhydrous, >= 99.9 %, inhibitor-free 1 * 1 L
Supplier: Merck
m-Xylene for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR® peroxide-free
Supplier: VWR Chemicals
Chloroform
Supplier: Roth Carl
Dodecamolybdophosphoric acid hydrate, Supelco®
Supplier: Merck
Dichloromethane 99.6% stabilised ACS
Supplier: Thermo Fisher Scientific
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific