(1R)-1-(5-bromo-2-fluoro-phenyl)ethanamine hydrochloride 1 * 1 g
Catalog # PHBLPBX0818-1-1G
Supplier: Pharmablock (USA)
(1R)-1-(5-bromo-2-fluoro-phenyl)ethanamine hydrochloride 1 * 1 g
Catalog # PHBLPBX0818-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99%, TECHNICAL
Supplier: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Glycerine (vegetable based), anhydrous, EMPROVE® EXPERT ACS, Ph. Eur., BP, USP, JP for biopharmaceutical production, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Methanol, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
n-Hexane ≥95%, TECHNICAL
Supplier: VWR Chemicals
Toluene ≥99%, GPR RECTAPUR® for pathology
Supplier: VWR Chemicals
1-Butanol ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals