(2R,3R)-2,3-dihydroxybutanedioic acid 2-{[(3aR,4S,6R,6aS)-6-amino-2,2-dimethyl-hexahydrocyclopenta[d][1,3]dioxol-4-yl]oxy}ethan-1-o l 1 * 5 g
Catalog # PHBLPBWX1001-1-5G
Supplier: Pharmablock (USA)
(2R,3R)-2,3-dihydroxybutanedioic acid 2-{[(3aR,4S,6R,6aS)-6-amino-2,2-dimethyl-hexahydrocyclopenta[d][1,3]dioxol-4-yl]oxy}ethan-1-o l 1 * 5 g
Catalog # PHBLPBWX1001-1-5G
Supplier: Pharmablock (USA)
Frequently Bought Together
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Solvent ASTM D2896, Chlorobenzene/Acetic acid-mixture (2/1 v/v), TBN solvent, VOLUSOL®
Supplier: VWR Chemicals
4-Methyl-2-pentanone ≥99%
Supplier: Thermo Fisher Scientific
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
1,4-Dioxane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
Supplier: Thermo Fisher Scientific
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
Methyl ethyl ketone ≥99%
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers) for histology
Supplier: Honeywell Chemicals
n-Hexane
Supplier: Roth Carl
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Chlorobenzene 99.9% for HPLC
Supplier: Thermo Fisher Scientific