(1S,3S,4S,5R)-2-tert-butoxycarbonyl-5-hydroxy-2-azabicyclo[2.2.2]octane-3-carboxylic acid 1 * 1 g
Catalog # PHBLPBWP393-1G
Supplier: Pharmablock (USA)
(1S,3S,4S,5R)-2-tert-butoxycarbonyl-5-hydroxy-2-azabicyclo[2.2.2]octane-3-carboxylic acid 1 * 1 g
Catalog # PHBLPBWP393-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Dichloromethane, extra dry over Molecular Sieve 99.8% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide 99.8+%, extra pure
Supplier: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals