4-iodo-1-[trans-4-[(tert-butyldimethylsilyl)oxy]cyclohexyl]-1H-pyrazole 1 * 1 g
Catalog # PHBLPBWB8212-1G
Supplier: Pharmablock (USA)
4-iodo-1-[trans-4-[(tert-butyldimethylsilyl)oxy]cyclohexyl]-1H-pyrazole 1 * 1 g
Catalog # PHBLPBWB8212-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Water, distilled
Supplier: DISTRIBUTOR
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Triethylamine ≥99.6%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol, SupraSolv® for GC-MS
Supplier: Merck
N,N-Dimethylformamide, anhydrous (max. 0.005% H₂O) ≥99.8%
Supplier: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals