(1R)-2,2,2-trideuterio-1-[3-(difluoromethyl)-2-fluoro-phenyl]ethanamine hydrochloride 1 * 1 g
Catalog # PHBLPBUE815-1-1G
Supplier: Pharmablock (USA)
(1R)-2,2,2-trideuterio-1-[3-(difluoromethyl)-2-fluoro-phenyl]ethanamine hydrochloride 1 * 1 g
Catalog # PHBLPBUE815-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
n-Heptane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
n-Hexane ≥99%, PESTINORM® Supra Trace for organic trace analysis
Supplier: VWR Chemicals
Ethanol absolute, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Ethyl acetate, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Methanol ≥98.5%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Diethyl ether stabilised, TECHNICAL, rectified
Supplier: VWR Chemicals
Methanol, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor