[2-fluoro-5-(trifluoromethoxy)phenyl]methanamine hydrochloride 1 * 1 g
Catalog # PHBLPBU9447-1-1G
Supplier: Pharmablock (USA)
[2-fluoro-5-(trifluoromethoxy)phenyl]methanamine hydrochloride 1 * 1 g
Catalog # PHBLPBU9447-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for UHPLC-MS
Supplier: VWR Chemicals
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
(±)-2-Methyltetrahydrofuran ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
Supplier: Merck
2-Propanol 70% ≥70% (v/v), GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
N-Methyl-2-pyrrolidone (NMP) ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Trifluoroacetic acid, Sigma-Aldrich®
Supplier: Merck
Tetrahydrofuran, EMPLURA®, extra pure, Supelco®
Supplier: Merck