benzyl 4-[(3S)-4-tert-butoxycarbonyl-3-(cyanomethyl)piperazin-1-yl]-2-methylsulfanyl-6,8-dihydro-5H-pyrido[3,4-d]pyrimidine-7-carbox ylate 1 * 1 g
Catalog # PHBLPBU7643-1G
Supplier: Pharmablock (USA)
benzyl 4-[(3S)-4-tert-butoxycarbonyl-3-(cyanomethyl)piperazin-1-yl]-2-methylsulfanyl-6,8-dihydro-5H-pyrido[3,4-d]pyrimidine-7-carbox ylate 1 * 1 g
Catalog # PHBLPBU7643-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
n-Nonane ≥95.0%, PESTINORM®
Supplier: VWR Chemicals
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Butyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
Petroleum spirit, 80…110 °C
Supplier: Roth Carl
Ethanol absolute, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Methanol with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Heptane ∼ 99% (n-heptane) for HPLC
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99.8% (by GC) stabilised, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND