1-[2-hydroxy-5-(trifluoromethyl)phenyl]ethanone 1 * 1 g
Catalog # PHBLPBU7046-1G
Supplier: Pharmablock (USA)
1-[2-hydroxy-5-(trifluoromethyl)phenyl]ethanone 1 * 1 g
Catalog # PHBLPBU7046-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
N-Hydroxysuccinimide for synthesis, Sigma-Aldrich®
Supplier: Merck
Water, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Petroleum spirit, 35…60 °C ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol absolute (untaxed) ≥99.8% (by GC), Puriss., Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Pentanol 99+% ACS
Supplier: Thermo Fisher Scientific
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
Dichloromethane 99.9% stabilised, AcroSeal® for biochemistry
Supplier: Thermo Fisher Scientific
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Dodecyltrimethylammonium bromide, Supelco®
Supplier: Merck
1,5-Diaminopentane 98%
Supplier: Thermo Fisher Scientific