4-chloro-8-isopropyl-2-methylsulfanyl-pyrazolo[1,5-a][1,3,5]triazine 1 * 1 g
Catalog # PHBLPBU7041-1G
Supplier: Pharmablock (USA)
4-chloro-8-isopropyl-2-methylsulfanyl-pyrazolo[1,5-a][1,3,5]triazine 1 * 1 g
Catalog # PHBLPBU7041-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethanol absolute ≥99.5%, EMPLURA®, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
tert-Butanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol 70% ≥70% (v/v), GPR RECTAPUR®
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Methanol, J.T.Baker®
Supplier: Avantor