methyl (3S,6S,8aS)-6-(tert-butoxycarbonylamino)-5-oxo-2,3,6,7,8,8a-hexahydro-1H-indolizine-3-carboxylate 1 * 1 g
Catalog # PHBLPBU7039-1G
Supplier: Pharmablock (USA)
methyl (3S,6S,8aS)-6-(tert-butoxycarbonylamino)-5-oxo-2,3,6,7,8,8a-hexahydro-1H-indolizine-3-carboxylate 1 * 1 g
Catalog # PHBLPBU7039-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
tert-Butyl methyl ether anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
1,2-Dichloroethane ≥99.8%, ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
Methanol ≥99.9% ACS, Spectrophotometric Grade for spectrophotometry, Riedel-de Haën™
Supplier: Honeywell Chemicals
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
n-Pentane, anhydrous 99+%, extra pure
Supplier: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
n-Hexane
Supplier: Roth Carl
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals
Ethyl acetate, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Ethoxyethanol 99%, Reagent Grade
Supplier: Honeywell Chemicals