(2S,4S)-N-[(1S)-1-(hydroxymethyl)-2,2-dimethyl-propyl]-9-(4-oxidopyrazin-4-ium-2-yl)-8,9-diazatricyclo[4.3.0.0]nona-1(6),7-diene-7 -carboxamide 1 * 1 g
Catalog # PHBLPBU6844-1G
Supplier: Pharmablock (USA)
(2S,4S)-N-[(1S)-1-(hydroxymethyl)-2,2-dimethyl-propyl]-9-(4-oxidopyrazin-4-ium-2-yl)-8,9-diazatricyclo[4.3.0.0]nona-1(6),7-diene-7 -carboxamide 1 * 1 g
Catalog # PHBLPBU6844-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals