[2-(difluoromethyl)-4-fluoro-phenyl]hydrazine hydrochloride 1 * 1 g
PHBLPBU5777-1-1G
[2-(difluoromethyl)-4-fluoro-phenyl]hydrazine hydrochloride 1 * 1 g
PHBLPBU5777-1-1G
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
n-Pentane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Toluene ≥98%, TECHNICAL
: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
: VWR Chemicals