[4-fluoro-2-(trifluoromethyl)phenyl]hydrazine hydrochloride 1 * 1 g
PHBLPBU5775-1-1G
[4-fluoro-2-(trifluoromethyl)phenyl]hydrazine hydrochloride 1 * 1 g
PHBLPBU5775-1-1G
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals