5-ethyl-2-fluoro-4-[4-fluoro-3-(4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridin-2-yl)-1H-indazol-6-yl]phenol dihydrochloride 1 * 1 g
Catalog # PHBLPBU5185-1-1G
Supplier: Pharmablock (USA)
5-ethyl-2-fluoro-4-[4-fluoro-3-(4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridin-2-yl)-1H-indazol-6-yl]phenol dihydrochloride 1 * 1 g
Catalog # PHBLPBU5185-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
2-Propanol ≥70% (v/v), SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Dichloromethane stabilised, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
tert-Butyl methyl ether anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Paraffin Oil heavy, liquid, Sigma-Aldrich®
Supplier: Merck
Ethanol (untaxed) ~96%, Puriss. p.a. ACS, without additive prima fine spirit, F15 (not denatured), Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.8% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Acetone ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene 99.5% ACS for spectroscopy
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexanone, Sigma-Aldrich®
Supplier: Merck
Ethanol (taxed) 96.0-97.2%, Puriss., meets analytical specification of BP, Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals