(3S,3aS,6aR)-2-tert-butoxycarbonyl-3,3a,4,5,6,6a-hexahydro-1H-cyclopenta[c]pyrrole-3-carboxylic acid 1 * 1 g
Catalog # PHBLPBU4947-1G
Supplier: Pharmablock (USA)
(3S,3aS,6aR)-2-tert-butoxycarbonyl-3,3a,4,5,6,6a-hexahydro-1H-cyclopenta[c]pyrrole-3-carboxylic acid 1 * 1 g
Catalog # PHBLPBU4947-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
m-Xylene, extra dry 99%, AcroSeal®
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide ≥99.9% (by GC), CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% for LC-MS
Supplier: Thermo Fisher Scientific
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
n-Pentane 98%, Reagent Grade
Supplier: Honeywell Chemicals
Kerosene, Purum
Supplier: Honeywell Chemicals
Ethanolamine ACS
Supplier: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for HPLC
Supplier: Thermo Fisher Scientific
1,3-Dioxolane 99.5+% stabilised, pure
Supplier: Thermo Fisher Scientific