tert-butyl 4-[3-(methanesulfonyloxy)propyl]piperazine-1-carboxylate hydrochloride 1 * 10 g
Catalog # PHBLU12235-01-10G
Supplier: Pharmablock (USA)
tert-butyl 4-[3-(methanesulfonyloxy)propyl]piperazine-1-carboxylate hydrochloride 1 * 10 g
Catalog # PHBLU12235-01-10G
Supplier: Pharmablock (USA)
Frequently Bought Together
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Pyrogallol for synthesis, Sigma-Aldrich®
Supplier: Merck
1,6-Dibromohexane for synthesis, Sigma-Aldrich®
Supplier: Merck
Xylene (mixture of isomers), extra dry 98+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.5%, Reagent Grade
Supplier: Honeywell Chemicals
1,4-Dioxane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
1,4-Dioxane ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Butylboronic acid, Supelco®
Supplier: Merck
Acetone ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals