18-[[(1S)-1-carboxy-4-[2-[2-[2-[2-[2-[2-(2,5-dioxopyrrolidin-1-yl)oxy-2-oxo-ethoxy]ethoxy]ethylamino]-2-oxo-ethoxy]ethoxy]ethylamino ]-4-oxo-butyl]amino]-18-oxo-octadecanoic acid 1 * 1 g
Catalog # PHBLPBU1071-1G
Supplier: Pharmablock (USA)
18-[[(1S)-1-carboxy-4-[2-[2-[2-[2-[2-[2-(2,5-dioxopyrrolidin-1-yl)oxy-2-oxo-ethoxy]ethoxy]ethylamino]-2-oxo-ethoxy]ethoxy]ethylamino ]-4-oxo-butyl]amino]-18-oxo-octadecanoic acid 1 * 1 g
Catalog # PHBLPBU1071-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals