tert-butyl 3-isopropyl-1,4,6,7-tetrahydropyrazolo[4,3-c]pyridine-5-carboxylate 1 * 1 g
Catalog # PHBLPBU1019-1G
Supplier: Pharmablock (USA)
tert-butyl 3-isopropyl-1,4,6,7-tetrahydropyrazolo[4,3-c]pyridine-5-carboxylate 1 * 1 g
Catalog # PHBLPBU1019-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Aniline, EMSURE® for analysis, Supelco®
Supplier: Merck
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Toluene ≥99.9% (by GC), SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
2,2,4-Trimethylpentane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Ethyl acetate ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
Glycerine (glycerol) USP, Ph. Eur., BP
Supplier: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals