(2S)-2-amino-2-(1-methylpyrazol-3-yl)ethanol dihydrochloride 1 * 1 g
Catalog # PHBLPBU0805-1-1G
Supplier: Pharmablock (USA)
(2S)-2-amino-2-(1-methylpyrazol-3-yl)ethanol dihydrochloride 1 * 1 g
Catalog # PHBLPBU0805-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Formamide, EMSURE® for analysis, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Acetone, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
2-Propanol 70% ≥70% (v/v), GPR RECTAPUR®
Supplier: VWR Chemicals
1,4-Dioxane ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
n-Hexane ≥99%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
1-Propanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals