methyl (2R)-2-[(2-chloro-5-nitro-pyrimidin-4-yl)-isopropyl-amino]butanoate 1 * 1 g
Catalog # PHBLPBTYC2126-1G
Supplier: Pharmablock (USA)
methyl (2R)-2-[(2-chloro-5-nitro-pyrimidin-4-yl)-isopropyl-amino]butanoate 1 * 1 g
Catalog # PHBLPBTYC2126-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
4-Aminobenzoic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Cyclohexane anhydrous, 99.5 % 1 * 1 L
Supplier: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Butyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
N-HEPTANE 95% CHROMAR HPLC | 2,5 DM3 1 * 2,5 L
Supplier: MACRON AVANTOR BRAND
ISE standards
Supplier: Hach
Dichloromethane, anhydrous ≥99.7% stabilised
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Pentane reagent grade, 98% 1 * 1 L
Supplier: Merck
2-Propanol 99.5% for HPLC
Supplier: Thermo Fisher Scientific
Iodomethane ≥99.5% stabilised
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals