(2R,3R)-2,3-bis(benzoyloxy)butanedioic acid (4S)-4-(3-bromophenyl)-6,8-dichloro-2-methyl-1,2,3,4-tetrahydroisoquinoline 1 * 1 g
Catalog # PHBLPBTT00159-2-1G
Supplier: Pharmablock (USA)
(2R,3R)-2,3-bis(benzoyloxy)butanedioic acid (4S)-4-(3-bromophenyl)-6,8-dichloro-2-methyl-1,2,3,4-tetrahydroisoquinoline 1 * 1 g
Catalog # PHBLPBTT00159-2-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
tert-Butyl methyl ether anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
METHANOL SUITABLE FOR HPLC >99.9% 1 * 2,5 L
Supplier: Merck
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Toluene 99.5% ACS for spectroscopy
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
5L Distilled Water 1 * 5 L
Supplier: REAGENT
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
TERT. BUTANOL FOR HPLC 99,5% 1 * 1 L
Supplier: Merck
1,2-Dichlorobenzene 99+% for HPLC
Supplier: Thermo Fisher Scientific