(4S)-4-(3-bromophenyl)-6,8-dichloro-2-methyl-3,4-dihydro-1H-isoquinoline 1 * 1 g
Catalog # PHBLPBTT00159-1G
Supplier: Pharmablock (USA)
(4S)-4-(3-bromophenyl)-6,8-dichloro-2-methyl-3,4-dihydro-1H-isoquinoline 1 * 1 g
Catalog # PHBLPBTT00159-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
n-Pentane ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
Pentane anhydrous, 99 % 1 * 1 L
Supplier: Merck
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol 99.8%, ECD tested for pesticide analysis for residue analysis
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Cyclohexane
Supplier: Roth Carl
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
5L Distilled Water 1 * 5 L
Supplier: REAGENT
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals