N-[(2S,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-methyl-2-oxo-chromen-7-yl)oxy-tetrahydropyran-3-yl]acetamide 1 * 1 g
Catalog # PHBLPBTS042-1G
Supplier: Pharmablock (USA)
N-[(2S,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-methyl-2-oxo-chromen-7-yl)oxy-tetrahydropyran-3-yl]acetamide 1 * 1 g
Catalog # PHBLPBTS042-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
5L Distilled Water 1 * 5 L
Supplier: REAGENT
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
TERT. BUTANOL FOR HPLC 99,5% 1 * 1 L
Supplier: Merck
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
n-Hexane
Supplier: Roth Carl
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
Rathburn Tetrahydrofluran HPLC 1 * 2,5 L
Supplier: REAGECON