[(2R,3R,4S,5R,6R)-3,4,5-triacetoxy-6-fluoro-tetrahydropyran-2-yl]methyl acetate 1 * 1 g
Catalog # PHBLPBTS017-1G
Supplier: Pharmablock (USA)
[(2R,3R,4S,5R,6R)-3,4,5-triacetoxy-6-fluoro-tetrahydropyran-2-yl]methyl acetate 1 * 1 g
Catalog # PHBLPBTS017-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
5L Distilled Water 1 * 5 L
Supplier: REAGENT
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol 99.5% for HPLC
Supplier: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
2-Propanol, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Acetone ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Hexane (mixture of isomers) 95+% for spectroscopy
Supplier: Thermo Fisher Scientific