(2S,3R,4S,5S,6R)-2-(2-chloro-4-nitro-phenoxy)-6-(hydroxymethyl)tetrahydropyran-3,4,5-triol 1 * 1 g
Catalog # PHBLPBTS012-1G
Supplier: Pharmablock (USA)
(2S,3R,4S,5S,6R)-2-(2-chloro-4-nitro-phenoxy)-6-(hydroxymethyl)tetrahydropyran-3,4,5-triol 1 * 1 g
Catalog # PHBLPBTS012-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Butyl acetate ≥99.5% (by GC), EMSURE® for analysis, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol, anhydrous (max. 0.003% H₂O) analytical reagent
Supplier: Merck
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Heptane (mixture of isomers), TECHNICAL
Supplier: VWR Chemicals
Acetonitrile, Prepsolv® for preparative chromatography
Supplier: Merck
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Pyridine, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
Pyridine ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1-Propanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Chloroform ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck