2-[6-(trifluoromethoxy)-1H-indol-3-yl]ethan-1-amine hydrochloride 1 * 1 g
Catalog # PHBLPBTKB0230-1-1G
Supplier: Pharmablock (USA)
2-[6-(trifluoromethoxy)-1H-indol-3-yl]ethan-1-amine hydrochloride 1 * 1 g
Catalog # PHBLPBTKB0230-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
1,4-Dioxane stabilised, EMPLURA®, Supelco®
Supplier: Merck
Ethyl acetate ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Methylbutane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
Toluene : 2-Propanol : Water (49,5 : 50 : 0,5) % vol., VOLUSOL® titration solvent
Supplier: VWR Chemicals
Diethyl ether, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Methyl ethyl ketone ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥99.9% (by GC) stabilised, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
2-Propanol, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
5L Distilled Water 1 * 5 L
Supplier: REAGENT
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Benzyl alcohol ≥99%
Supplier: Thermo Fisher Scientific