2-[5-(trifluoromethoxy)-1H-indol-3-yl]ethan-1-amine hydrochloride 1 * 1 g
Catalog # PHBLPBTKB0229-1-1G
Supplier: Pharmablock (USA)
2-[5-(trifluoromethoxy)-1H-indol-3-yl]ethan-1-amine hydrochloride 1 * 1 g
Catalog # PHBLPBTKB0229-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
2-Propanol, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane for gas chromatography, Burdick & Jackson™
Supplier: Honeywell Chemicals
2-Butoxyethanol 99%, extra pure
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Pentane, anhydrous 99+%, extra pure
Supplier: Thermo Fisher Scientific
2-Propanol 99.5% for HPLC
Supplier: Thermo Fisher Scientific
Glycerine, Hi-LR™
Supplier: HIMEDIA
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
1-Pentanol ≥98%
Supplier: Thermo Fisher Scientific
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
Acetone, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
5L Distilled Water 1 * 5 L
Supplier: REAGENT
Xylene (mixture of isomers), extra dry 98+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific