2-{8-oxa-3-azabicyclo[3.2.1]octan-3-yl}ethan-1-amine hydrochloride 1 * 1 g
Catalog # PHBLPBTEN154521-1G
Supplier: Pharmablock (USA)
2-{8-oxa-3-azabicyclo[3.2.1]octan-3-yl}ethan-1-amine hydrochloride 1 * 1 g
Catalog # PHBLPBTEN154521-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethyl acetate, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
N,N-Dimethylacetamide ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
n-Heptane ≥99.0%, PESTINORM® Supra Trace for organic trace analysis
Supplier: VWR Chemicals
Acetone, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
Methanol ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
n-Hexane ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ethylene glycol 99.5% for analysis
Supplier: Thermo Fisher Scientific
Ethanol 80% vol, Molecular Biology Grade
Supplier: VWR Chemicals
Ethanol (untaxed) ~96%, Puriss. p.a. ACS, without additive prima fine spirit, F15 (not denatured), Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Mineral oil (petroleum) for IR spectrophotometry, suitable for nujol mull preparations
Supplier: Thermo Fisher Scientific
Acetone, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific