methyl 2,6-dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 1 * 1 g
Catalog # PHBLPBTD233-1G
Supplier: Pharmablock (USA)
methyl 2,6-dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 1 * 1 g
Catalog # PHBLPBTD233-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
n-Pentane ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
(±)-2-Butanol, EMSURE® for analysis, Supelco®
Supplier: Merck
Acetone, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Dimethyl Sulfoxide EMPROVE EXPERT Ph Eur,USP 1 * 1 L
Supplier: MERCK PRODUCTION CHEMICALS
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethylene glycol ≥98%, TECHNICAL
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.8% for peptide synthesis
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1-Butanol ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Triethylamine, Sigma-Aldrich®
Supplier: Merck
Water for LC-MS 1 * 2,5 L
Supplier: PanReac AppliChem
2,2,4-Trimethylpentane 99+%, pure
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide ≥99%
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.5%, Reagent Grade
Supplier: Honeywell Chemicals