1-[(2R,4S,5R)-5-[[tert-butyl(dimethyl)silyl]oxymethyl]-4-hydroxy-tetrahydrofuran-2-yl]-5-methyl-pyrimidine-2,4-dione 1 * 1 g
Catalog # PHBLPBTB717-1G
Supplier: Pharmablock (USA)
1-[(2R,4S,5R)-5-[[tert-butyl(dimethyl)silyl]oxymethyl]-4-hydroxy-tetrahydrofuran-2-yl]-5-methyl-pyrimidine-2,4-dione 1 * 1 g
Catalog # PHBLPBTB717-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
1-Butanol ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Cyclopentyl methyl ether (CPME), EMPLURA®
Supplier: Merck
tert-Butyl methyl ether, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Methanol ≥99.9%, HiPerSolv CHROMANORM®, PFAS grade for LC-MS
Supplier: VWR Chemicals
Chloroform ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
tert-Butanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Petroleum spirit, 100…120 °C, AnalaR NORMAPUR® ACS analytical reagent
Supplier: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Heptane, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dichloromethane, anhydrous ≥99.7% stabilised
Supplier: Thermo Fisher Scientific
2-ETHOXYETHANOL, REAGENTPLUS, 99%. 1 * 1 Each
Supplier: Merck
n-Heptane 95%
Supplier: Thermo Fisher Scientific
EX-CELL Advanced CHO Fed-batch Medium 1 * 1.000 mL
Supplier: Merck