bis(1-adamantyl)-(2-morpholinophenyl)phosphane methylsulfonyloxy-(2-phenylanilino)palladium 1 * 1 g
Catalog # PHBLPBTA272-1G
Supplier: Pharmablock (USA)
bis(1-adamantyl)-(2-morpholinophenyl)phosphane methylsulfonyloxy-(2-phenylanilino)palladium 1 * 1 g
Catalog # PHBLPBTA272-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Cyclohexanone, EMPLURA®, extra pure, Supelco®
Supplier: Merck
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
2-Nitrophenylhydrazine, moistened 30% with water 97%
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
n-Hexane
Supplier: Roth Carl
Glycerine ≥83.5-88.5% (w/w) in aqueous solution, pure
Supplier: Thermo Fisher Scientific
ALCH, CHROMASOLV™ for HPLC (denatured with 5% isopropanol and 5% methanol), Riedel-de Haën™
Supplier: Honeywell Chemicals
5L Distilled Water 1 * 5 L
Supplier: REAGENT
Benzonitrile for HPLC
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals