3-[[(2R,3R,4R,5R)-2-[[bis(4-methoxyphenyl)-phenyl-methoxy]methyl]-5-(2,4-dioxopyrimidin-1-yl)-4-(2-methoxyethoxy)tetrahydrofuran-3-y l]oxy-(diisopropylamino)phosphanyl]oxypropanenitrile 1 * 1 g
Catalog # PHBLPBT8306-1G
Supplier: Pharmablock (USA)
3-[[(2R,3R,4R,5R)-2-[[bis(4-methoxyphenyl)-phenyl-methoxy]methyl]-5-(2,4-dioxopyrimidin-1-yl)-4-(2-methoxyethoxy)tetrahydrofuran-3-y l]oxy-(diisopropylamino)phosphanyl]oxypropanenitrile 1 * 1 g
Catalog # PHBLPBT8306-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥98.5%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99%, TECHNICAL
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals