(1R,2S,5R)-2-isopropyl-5-methyl-cyclohexanecarboxylic acid 1 * 1 g
Catalog # PHBLPBT8000-1G
Supplier: Pharmablock (USA)
(1R,2S,5R)-2-isopropyl-5-methyl-cyclohexanecarboxylic acid 1 * 1 g
Catalog # PHBLPBT8000-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Benzoic acid methyl ester
Supplier: Merck
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Ethanol absolute, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
n-Heptane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
Petroleum spirit, 100…120 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
1-Butanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane 95+%, extra pure
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.8% ACS for spectroscopy
Supplier: Thermo Fisher Scientific