ditert-butyl-[6-methoxy-3-methyl-2-(2,4,6-triisopropylphenyl)phenyl]phosphane 1 * 1 g
Catalog # PHBLPBT4676-1G
Supplier: Pharmablock (USA)
ditert-butyl-[6-methoxy-3-methyl-2-(2,4,6-triisopropylphenyl)phenyl]phosphane 1 * 1 g
Catalog # PHBLPBT4676-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Trifluoroacetic acid ≥99.8% (by anhydrous basis), SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane ≥95%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
N,N-Dimethylformamide 99.0-100.00%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Triethylamine ≥99.6%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethyl acetate, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Acetonitrile with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Trichloroacetic Acid (BP, Ph. Eur.) pure, pharma grade 1 * 5 kg
Supplier: PanReac AppliChem
Methanol ≥99.8% for LC-MS
Supplier: Thermo Fisher Scientific