(2R,3R,4S,5R)-2-(4-chloropyrrolo[2,3-d]pyrimidin-7-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol 1 * 1 g
PHBLPBT4218-1G
(2R,3R,4S,5R)-2-(4-chloropyrrolo[2,3-d]pyrimidin-7-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol 1 * 1 g
PHBLPBT4218-1G
Frequently Bought Together
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
: PanReac AppliChem
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
tert-Butyl methyl ether, extra dry over Molecular Sieve 99+%, AcroSeal®
: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
: PanReac AppliChem
Dibutyl ether, extra dry 99+%, AcroSeal®
: Thermo Fisher Scientific
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
: PanReac AppliChem
n-Pentane 98%, Reagent Grade
: Honeywell Chemicals
Cyclohexane, extra dry 99.5%, AcroSeal®
: Thermo Fisher Scientific