3,5-dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 1 * 1 g
Catalog # PHBLPBT2608-1G
Supplier: Pharmablock (USA)
3,5-dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 1 * 1 g
Catalog # PHBLPBT2608-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethyl acetate, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Sulphur trioxide pyridine complex for synthesis
Supplier: Merck
Dimethyl sulphoxide, dehydrated (max. 0.01% H₂O) ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Chloroform 99.0-99.6% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Turpentine oil
Supplier: Roth Carl
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Pentane reagent grade, 98% 1 * 1 L
Supplier: Merck
n-Octane ≥98%
Supplier: Thermo Fisher Scientific
Acetic acid 99-100%
Supplier: Roth Carl
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Chlorobenzene ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Vaseline Oil (USP, BP, Ph. Eur.) pure, pharma grade 1 * 1.000 mL
Supplier: PanReac AppliChem
Methyl ethyl ketone ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals