(R)-(6-methoxy-4-quinolyl)-[(1S,2S,4S,5R)-5-vinylquinuclidin-2-yl]methanol 1 * 25 g
Catalog # PHBLPBT1367-25G
Supplier: Pharmablock (USA)
(R)-(6-methoxy-4-quinolyl)-[(1S,2S,4S,5R)-5-vinylquinuclidin-2-yl]methanol 1 * 25 g
Catalog # PHBLPBT1367-25G
Supplier: Pharmablock (USA)
Frequently Bought Together
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Methanol ≥98.5%, TECHNICAL
Supplier: VWR Chemicals
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane stabilised, EMPLURA®
Supplier: Merck
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Pyridine ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, TraceSELECT™ for trace analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99.5%, Environmental Grade
Supplier: Thermo Fisher Scientific
2-Aminoanthracene 94%
Supplier: Thermo Fisher Scientific
n-Pentane, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Acetone ≥99.5% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Triethylamine, Sigma-Aldrich®
Supplier: Merck