tert-butyl N-[(1S,3S)-1-[(2S,4S)-4-isopropyl-5-oxo-tetrahydrofuran-2-yl]-3-[[4-methoxy-3-(3-methoxypropoxy)phenyl]methyl]-4-methyl-p entyl]carbamate 1 * 1 g
Catalog # PHBLPBT0975-1G
Supplier: Pharmablock (USA)
tert-butyl N-[(1S,3S)-1-[(2S,4S)-4-isopropyl-5-oxo-tetrahydrofuran-2-yl]-3-[[4-methoxy-3-(3-methoxypropoxy)phenyl]methyl]-4-methyl-p entyl]carbamate 1 * 1 g
Catalog # PHBLPBT0975-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
n-Heptane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, EMPLURA®, Supelco®
Supplier: Merck
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals