[1,3-bis(2,4,6-trimethylphenyl)imidazolidin-2-ylidene]-dichloro-[(2-isopropoxyphenyl)methylene]ruthenium 1 * 1 g
Catalog # PHBLPBT0888-1G
Supplier: Pharmablock (USA)
[1,3-bis(2,4,6-trimethylphenyl)imidazolidin-2-ylidene]-dichloro-[(2-isopropoxyphenyl)methylene]ruthenium 1 * 1 g
Catalog # PHBLPBT0888-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
1,3-Dioxolane ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
n-Hexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Cyclohexane ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Trichloroethylene 99.6% stabilised ACS
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylformamide ≥99.8% ACS for spectroscopy
Supplier: Thermo Fisher Scientific
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals