(1R,2S,5R)-2-isopropyl-5-methyl-cyclohexanol 1 * 10 g
Catalog # PHBLPBT0309-10G
Supplier: Pharmablock (USA)
(1R,2S,5R)-2-isopropyl-5-methyl-cyclohexanol 1 * 10 g
Catalog # PHBLPBT0309-10G
Supplier: Pharmablock (USA)
Frequently Bought Together
1-Butanol ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
o-Xylene anhydrous, 97 % 1 * 1 L
Supplier: Merck
m-Xylene for synthesis, Sigma-Aldrich®
Supplier: Merck
n-Octane 97%, pure
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
1-Propanol ACS for spectroscopy
Supplier: Thermo Fisher Scientific
1,3-Propanediol ≥99%
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) ≥99%
Supplier: Thermo Fisher Scientific
Tetrahydrofuran stabilized with 300 ppm of BHT for analysis, ACS 1 * 1.000 mL
Supplier: PanReac AppliChem
Chloroform ≥99.8% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Ethylene glycol, Sigma-Aldrich®
Supplier: Merck
1,3-Dihydroxynaphthalene, Supelco®
Supplier: Merck
1,2-Dichloroethane 99+%, pure
Supplier: Thermo Fisher Scientific
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
n-Pentane 98%, Reagent Grade
Supplier: Honeywell Chemicals