3-isopropyl-5-methylsulfanyl-1,6-dihydropyrazolo[4,3-d]pyrimidin-7-one 1 * 1 g
PHBLPBSL027-1G
3-isopropyl-5-methylsulfanyl-1,6-dihydropyrazolo[4,3-d]pyrimidin-7-one 1 * 1 g
PHBLPBSL027-1G
Frequently Bought Together
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals