(1R)-3-methyl-1-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)butan-1-amine hydrochloride 1 * 1 g
Catalog # PHBLPBSL0012-1-1G
Supplier: Pharmablock (USA)
(1R)-3-methyl-1-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)butan-1-amine hydrochloride 1 * 1 g
Catalog # PHBLPBSL0012-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
(±)-2-Methyl-2,4-pentanediol for synthesis, Sigma-Aldrich®
Supplier: Merck
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Supplier: PanReac AppliChem
Bromoform 99+% stabilised
Supplier: Thermo Fisher Scientific
Glycerine ≥83.5-88.5% (w/w) in aqueous solution, pure
Supplier: Thermo Fisher Scientific
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Cyclohexane ≥99.5% (by GC), ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Isobutanol, Sigma-Aldrich®
Supplier: Merck
Diethyl ether, anhydrous 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
1-Butanol ≥99.4% ACS
Supplier: Thermo Fisher Scientific
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
Methanol with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals