[(2R,3R,4S,5R,6R)-3,4,5-tris(acetyloxy)-6-hydroxyoxan-2-yl]methyl acetate 1 * 1 g
Catalog # PHBLPBSB058-1G
Supplier: Pharmablock (USA)
[(2R,3R,4S,5R,6R)-3,4,5-tris(acetyloxy)-6-hydroxyoxan-2-yl]methyl acetate 1 * 1 g
Catalog # PHBLPBSB058-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetonitrile, anhydrous (max. 0.001% H₂O) ≥99.8%
Supplier: VWR Chemicals
Ethanol absolute, EMPROVE® EXPERT Ph. Eur., BP, ChP, JP, USP suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
N-(1-Naphthyl)ethylenediamine dihydrochloride, Supelco®
Supplier: Merck
N,N-Dimethylacetamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
Acetonitrile 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Tetrabutylammonium dihydrogen phosphate 99% for HPLC
Supplier: Thermo Fisher Scientific
n-Hexane
Supplier: Roth Carl
N-Methyl-2-pyrrolidone (NMP) 98%, pure
Supplier: Thermo Fisher Scientific
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
5L Distilled Water 1 * 5 L
Supplier: REAGENT
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers), J.T.Baker®
Supplier: Avantor