1-[(2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl]-5-methyl-2-thioxo-pyrimidin-4-one 1 * 1 g
Catalog # PHBLPBQS735-1G
Supplier: Pharmablock (USA)
1-[(2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl]-5-methyl-2-thioxo-pyrimidin-4-one 1 * 1 g
Catalog # PHBLPBQS735-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Pyridine, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
Ethanol absolute, SupraSolv® for GC-ECD/FID
Supplier: Merck
Ethylene glycol, EMPLURA®, Supelco®
Supplier: Merck
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
Pyridine, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Water, BAKER ANALYZED®, J.T.Baker®
Supplier: Avantor
1-Butanol for HPLC
Supplier: Thermo Fisher Scientific