3-[[(2R,3R,4R,5R)-2-[[bis(4-methoxyphenyl)-phenyl-methoxy]methyl]-4-(2-methoxyethoxy)-5-(5-methyl-2,4-dioxo-pyrimidin-1-yl)tetrahydr ofuran-3-yl]oxy-(diisopropylamino)phosphanyl]oxypropanenitrile 1 * 1 g
Catalog # PHBLPBQS707-1G
Supplier: Pharmablock (USA)
3-[[(2R,3R,4R,5R)-2-[[bis(4-methoxyphenyl)-phenyl-methoxy]methyl]-4-(2-methoxyethoxy)-5-(5-methyl-2,4-dioxo-pyrimidin-1-yl)tetrahydr ofuran-3-yl]oxy-(diisopropylamino)phosphanyl]oxypropanenitrile 1 * 1 g
Catalog # PHBLPBQS707-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, EMPLURA®, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals