N-{9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-9H-purin-6-yl}benzamide 1 * 10 g
Catalog # PHBLPBQN065-10G
Supplier: Pharmablock (USA)
N-{9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-9H-purin-6-yl}benzamide 1 * 10 g
Catalog # PHBLPBQN065-10G
Supplier: Pharmablock (USA)
Frequently Bought Together
Butyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butyl methyl ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.7% for HPLC
Supplier: Thermo Fisher Scientific
Methanol with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Dichloromethane ≥99.7% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Diisopropyl ether 98+% stabilised, pure
Supplier: Thermo Fisher Scientific
Acetone, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Hexane (mixture of isomers) 99% ACS
Supplier: Thermo Fisher Scientific